Merck
  • Intraductal fulvestrant for therapy of ERα-positive ductal carcinoma in situ of the breast: a preclinical study.
Intraductal fulvestrant for therapy of ERα-positive ductal carcinoma in situ of the breast: a preclinical study.
Carcinogenesis (2019-05-03)
Guannan Wang, Chuang Chen, Priya Pai, Preethi Korangath, Shengrong Sun, Vanessa F Merino, Jingping Yuan, Suping Li, Guangjun Nie, Vered Stearns, Saraswati Sukumar
ABSTRACT
Mammographic screening for breast cancer has led to increased detection of ductal carcinoma in situ (DCIS) and a reappraisal of the necessity of aggressive treatment with their attendant toxicities for a preneoplastic lesion. Fulvestrant, a selective estrogen receptor degrader, is very effective in the treatment of estrogen receptor positive (ER+) breast cancer, but delivery by the painful intramuscular (i.m) route is limiting. We hypothesized that intraductal (i.duc) administration of fulvestrant will provide a direct, safe and effective treatment for DCIS. Mice bearing mammary ductal xenografts of ER+, luciferase-tagged MCF-7 breast cancer cells were administered vehicle or fulvestrant i.m or i.duc. I.duc MCF-7-luc tumors in mice treated with fulvestrant i.duc or i.m grew significantly slower than vehicle control. Whole mount analysis and histopathology showed that i.duc fulvestrant achieved significantly larger cancer-free areas. Western blot analysis showed reduced levels of estrogen receptor alpha (ERα) and its downstream targets, c-Myc and Cyclin D1, and increased levels of ERβ, which is known to inhibit ERα function. Immunohistochemical analysis of tumor sections showed that Ki67 and ERα protein levels decreased by 3-fold, and neoangiogenesis was inhibited by i.duc fulvestrant treatment. I.duc fulvestrant also reduced outgrowth of ERα+, autochthonous N-methyl-N-nitrosourea-induced mammary tumors in rats. Overall, we have shown that i.duc fulvestrant was significantly more effective than, or equivalent in action to i.m fulvestrant in two preclinical models of breast cancer. These studies provide evidence for a novel and safe route for fulvestrant therapy of DCIS and prevention of breast cancer. This preclinical study provides a strong basis for conducting clinical trials for DCIS and early breast cancer.
Sigma-Aldrich
Castor oil
Sigma-Aldrich
Carmine, powder, ≥42.0% (UV-vis)
Sigma-Aldrich
Fulvestrant, >98% (HPLC)
Sigma-Aldrich
Benzyl benzoate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture