Merck
  • SPC18 Expression Is an Independent Prognostic Indicator of Patients with Esophageal Squamous Cell Carcinoma.
SPC18 Expression Is an Independent Prognostic Indicator of Patients with Esophageal Squamous Cell Carcinoma.
Pathobiology : journal of immunopathology, molecular and cellular biology (2020-06-22)
Yuji Yamamoto, Naohide Oue, Ryuichi Asai, Narutaka Katsuya, Naohiro Uraoka, Naoya Sakamoto, Kazuhiro Sentani, Kazuaki Tanabe, Hideki Ohdan, Wataru Yasui
ABSTRACT
Esophageal cancer is the sixth most common malignancy worldwide. Signal peptidase complex 18 (SPC18) protein, which is encoded by the SEC11A gene, is one of the subunits of the signal peptidase complex and plays an important role in the secretion of proteins including transforming growth factor α (TGF-α). In this study, we investigated the significance of SPC18 expression in human esophageal squamous cell carcinoma (ESCC). SPC18 expression was examined by immunohistochemistry. RNA interference was used to inhibit SPC18 expression in ESCC cell lines. To examine cell viability, we performed 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assays. The effects of SPC18 inhibition on epidermal growth factor receptor (EGFR) signaling were analyzed by Western blot. In total, 46 (50%) of 92 ESCC cases were positive for SPC18. SPC18 staining was observed more frequently in stage II/III/IV cases than in stage I cases (p = 0.028). We found that SPC18 expression was significantly associated with increased cancer-specific mortality (p = 0.006, log-rank test). SPC18 expression was frequently found in EGFR-positive cases compared with EGFR-negative cases. Cell proliferation and EGFR signaling were inhibited by SPC18 knockdown. Specific inhibitors of SPC18 may be promising anticancer drugs for patients with ESCC.