Merck
  • Nitrogen acquisition from mineral-associated proteins by an ectomycorrhizal fungus.
Nitrogen acquisition from mineral-associated proteins by an ectomycorrhizal fungus.
The New phytologist (2020-04-13)
Tao Wang, Zhaomo Tian, Anders Tunlid, Per Persson
ABSTRACT
In nitrogen (N)-limited boreal forests, trees depend on the decomposing activity of their ectomycorrhizal (ECM) fungal symbionts to access soil N. A large fraction of this N exists as proteinaceous compounds associated with mineral particles. However, it is not known if ECM fungi can access these mineral-associated proteins; accordingly, possible acquisition mechanisms have not been investigated. With tightly controlled isotopic, spectroscopic, and chromatographic experiments, we quantified and analyzed the mechanisms of N acquisition from iron oxide mineral-associated proteins by Paxillus involutus, a widespread ECM fungus in boreal forests. The fungus acquired N from the mineral-associated proteins. The collective results indicated a proteolytic mechanism involving formation of the crucial enzyme-substrate complexes at the mineral surfaces. Hence, the enzymes hydrolyzed the mineral-associated proteins without initial desorption of the proteins. The proteolytic activity was suppressed by adsorption of proteases to the mineral particles. This process was counteracted by fungal secretion of mineral-surface-reactive compounds that decreased the protease-mineral interactions and thereby promoted the formation of enzyme-substrate complexes. The ability of ECM fungi to simultaneously generate extracellular proteases and surface-reactive metabolites suggests that they can play an important role in unlocking the large N pool of mineral-associated proteins to trees in boreal forests.
Sigma-Aldrich
Goethite, 30-63% Fe