Merck
  • Growth inhibitory activities of oxyprenylated and non-prenylated naturally occurring phenylpropanoids in cancer cell lines.
Growth inhibitory activities of oxyprenylated and non-prenylated naturally occurring phenylpropanoids in cancer cell lines.
Bioorganic & medicinal chemistry letters (2011-06-24)
Céline Bruyère, Salvatore Genovese, Benjamin Lallemand, Alexandra Ionescu-Motatu, Massimo Curini, Robert Kiss, Francesco Epifano
ABSTRACT
A series of 25 selected oxyprenylated natural phenylpropanoids were synthesized, and their growth inhibitory activities were evaluated in vitro together with 14 other commercially available non-alkylated compounds belonging to the same chemical series. The compounds were tested on six human cancer cell lines using MTT colorimetric assays. The data reveal that of the six chemical groups (G) studied, coumarins (G1), cinnamic and benzoic acids (G2), chalcones (G3), acetophenones (G4), anthraquinones (G5), and cinnamaldehydes and cinnamyl alcohols (G6), G2-related compounds displayed the weakest growth inhibitory activities in vitro, whereas G5-related compounds displayed the highest activities. Quantitative videomicroscopy analyses were then carried out on human U373 glioblastoma cells, which are characterized by various levels of resistance to different pro-apoptotic stimuli. These analyses revealed that compounds 20 (4,2',4'-trihydroxychalcone), and 30 and 31 (two cinnamaldehydes) were cytostatic and able to overcome the intrinsic resistance of U373 cancer cells to pro-apoptotic stimuli.
Sigma-Aldrich
Coniferyl alcohol, 98%
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Emodin, from Frangula bark, ≥90% (HPLC)
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Vanillic acid, 97%
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Vanillic acid, ≥97%, FG
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)