Merck
  • Shortwave-infrared meso-patterned imaging enables label-free mapping of tissue water and lipid content.
Shortwave-infrared meso-patterned imaging enables label-free mapping of tissue water and lipid content.
Nature communications (2020-10-25)
Yanyu Zhao, Anahita Pilvar, Anup Tank, Hannah Peterson, John Jiang, Jon C Aster, John Paul Dumas, Mark C Pierce, Darren Roblyer
ABSTRACT
Water and lipids are key participants in many biological processes, but there are few non-invasive methods that provide quantification of these components in vivo, and none that can isolate and quantify lipids in the blood. Here we develop a new imaging modality termed shortwave infrared meso-patterned imaging (SWIR-MPI) to provide label-free, non-contact, spatial mapping of water and lipid concentrations in tissue. The method utilizes patterned hyperspectral illumination to target chromophore absorption bands in the 900-1,300 nm wavelength range. We use SWIR-MPI to monitor clinically important physiological processes including edema, inflammation, and tumor lipid heterogeneity in preclinical models. We also show that SWIR-MPI can spatially map blood-lipids in humans, representing an example of non-invasive and contact-free measurements of in vivo blood lipids. Together, these results highlight the potential of SWIR-MPI to enable new capabilities in fundamental studies and clinical monitoring of major conditions including obesity, cancer, and cardiovascular disease.
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Nigrosin water soluble, For use as a biological stain
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%