- 5-Substituted [1]pyrindine derivatives with antiproliferative activity.
5-Substituted [1]pyrindine derivatives with antiproliferative activity.
European journal of medicinal chemistry (2009-12-09)
Stéphanie Kolb, Mary-Lorène Goddard, Ali Loukaci, Odile Mondésert, Bernard Ducommun, Emmanuelle Braud, Christiane Garbay
PMID19969400
ABSTRACT
We report herein the synthesis of 5-substituted [1]pyrindine derivatives and the evaluation of their antiproliferative properties on HeLa cells, a cervical carcinoma tumor cell line, and on the melanoma A2058 cell line. The most efficient compounds display cytotoxicity against tumor cells in the micromolar range but have interestingly no effect against the normal human fibroblasts CRL-2796. Generally, these pyrindines are active on both tumor cell lines. Compounds bearing large substituents with structural rigidity at position 5 such as phenyl-furyl show no inhibition of cell growth.
Product Description
A2058, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 91100402, human skin (lymph node metastasis), Epithelial