Merck
  • Effect of pH and urea on the proteins secondary structure at the water/air interface and in solution.
Effect of pH and urea on the proteins secondary structure at the water/air interface and in solution.
Journal of colloid and interface science (2021-02-02)
Tobias Guckeisen, Saman Hosseinpour, Wolfgang Peukert
ABSTRACT
The secondary structure of proteins affects their functionality and performance in physiological environments or industrial applications. Change of the solution pH or the presence of protein denaturants are the main chemical means that can alter the secondary structure of proteins or lead to protein denaturation. Since proteins in the bulk solution and those residing at the solution/air interface experience different local environments, their response to chemical denaturation can be different. We utilize circular dichroism and chiral/achiral sum frequency generation spectroscopy to study the secondary structure of selected proteins as a function of the solution pH or in the presence of 8 M urea in the bulk solution and at the solution/air interface, respectively. The liquid/air interface can enhance or decrease protein conformation stability. The change in the secondary structure of the surface adsorbed proteins in alkaline solutions occurs at pH values lower than those denaturing the studied proteins in the bulk solution. In contrast, while 8 M urea completely denatures the studied proteins in the bulk solution, the liquid/air interface prevents the urea-induced denaturation of the surface adsorbed proteins by limiting the access of urea to the hydrophobic side chains of proteins protruding to air.
Sigma-Aldrich
Catalase from bovine liver, Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, 2,000-5,000 units/mg protein
Sigma-Aldrich
α-Chymotrypsinogen A from bovine pancreas, essentially salt-free, lyophilized powder
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)