Merck
  • Synthesis, spectral analysis and in vitro microbiological evaluation of 3-(3-alkyl-2,6-diarylpiperin-4-ylidene)-2-thioxoimidazolidin-4-ones as a new class of antibacterial and antifungal agents.
Synthesis, spectral analysis and in vitro microbiological evaluation of 3-(3-alkyl-2,6-diarylpiperin-4-ylidene)-2-thioxoimidazolidin-4-ones as a new class of antibacterial and antifungal agents.
Bioorganic & medicinal chemistry letters (2009-12-17)
J Thanusu, V Kanagarajan, M Gopalakrishnan
ABSTRACT
In the present work, a new series of bis hybrid heterocycle comprising both piperidine and thiohydantoin nuclei together namely 3-(3-alkyl-2,6-diarylpiperin-4-ylidene)-2-thioxoimidazolidin-4-ones 46-60 was synthesized by the treatment of the respective thiosemicarbazones 31-45 with chloroethyl acetate and anhydrous sodium acetate in refluxing ethanol for 4h and were characterized by melting point, elemental analysis, MS, FT-IR, one-dimensional NMR ((1)H, D(2)O exchanged (1)H and (13)C), two dimensional HOMOCOSY and NOESY spectroscopic data. In addition, the title compounds were screened for their antimicrobial activities against a spectrum of clinically isolated microbial organisms. Compounds 47-50, 52-55 and 57-60 with fluoro, chloro, methoxy or methyl functions at the para position of phenyl rings attached to C-2 and C-6 carbons of piperidine moiety along with and without methyl substituent at position C-3 of the piperidine ring exerted potent biological activities against Staphylococcus aureus, beta-Hemolytic streptococcus, Vibrio cholerae, Escherichia coli, Pseudomonas aeruginosa, Aspergillus flavus, Candida albicans, Candida 6 and Candida 51 at a minimum inhibitory concentration.
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
Ciprofloxacin, VETRANAL®, analytical standard