Merck
  • Protective Activity of Aβ on Cell Cultures (PC12 and THP-1 after Differentiation) Preincubated with Lipopolysaccharide (LPS).
Protective Activity of Aβ on Cell Cultures (PC12 and THP-1 after Differentiation) Preincubated with Lipopolysaccharide (LPS).
Molecular neurobiology (2020-11-15)
Benita Wiatrak, Katarzyna Balon
ABSTRACT
Amyloid-β (Aβ), the influence of which is considered the pathomechanism of Alzheimer's disease, is also present in healthy people. The microbiome's impact is also taken into account, where bacterial lipopolysaccharide (LPS) activates inflammatory processes and stimulates microglia via TLRs. Molecules of bacterial origin can co-create senile plaques with Aβ. This study evaluated the activity of physiological Aβ concentrations on neuronal and microglial cells after preincubation with LPS. Two cell lines were used in the study: PC12 cells differentiated with NGF and THP-1 cells differentiated with phorbol 12-myristate 13-acetate (PMA). Cells were incubated with LPS at concentrations of 1-100 μM for 24 h and then with Aβ25-35 at a concentration of 0.001 μM or 1.0 μM for another 24 h. The viability of the culture and free oxygen radicals and the number of DNA strand breaks in both cell lines were evaluated. Additionally, for PC12 cells, neural features were assessed. Stimulation of repair processes in the presence of Aβ was observed for both studied cell lines. There was a decrease in free radical level and DNA damage number compared to control cultures (cells treated with LPS and without Aβ). The neurotrophic activity of Aβ was observed-the effect on neurites' growth even after the preincubation of PC12 cells with LPS. At the lowest concentration of LPS used, the increase in neurite length was about 50% greater than in the negative control. At low concentrations, Aβ has a protective effect on neuron-like PC12 cells pretreated with LPS.
Sigma-Aldrich
Amyloid β-Protein Fragment 25-35, ≥97% (HPLC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by phenol extraction