Merck
  • Prevalence of aac(6')-Ib-cr encoding a ciprofloxacin-modifying enzyme among Enterobacteriaceae blood isolates in Korea.
Prevalence of aac(6')-Ib-cr encoding a ciprofloxacin-modifying enzyme among Enterobacteriaceae blood isolates in Korea.
Antimicrobial agents and chemotherapy (2009-03-18)
Eun Sil Kim, Jin-Yong Jeong, Jae-Bum Jun, Sang-Ho Choi, Sang-Oh Lee, Mi-Na Kim, Jun Hee Woo, Yang Soo Kim
ABSTRACT
The aac(6')-Ib gene was detected in 86 of 555 (15.5%) Enterobacteriaceae isolates. Among these 86 aac(6')-Ib-positive isolates, 19 (22.0%) were positive for aac(6')-Ib-cr: 4 of 31 (12.9%) Enterobacter spp., 7 of 13 (53.8%) Escherichia coli isolates, and 8 of 42 (19.0%) Klebsiella pneumoniae isolates. There was a strong association between aac(6')-Ib-cr and OXA-1 and CTX-M-1 group beta-lactamase genes. One aac(6')-Ib-positive K. pneumoniae isolate carried both aac(6')-Ib-cr and qnrS.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Nalidixic acid, ≥98%
Supelco
Norfloxacin, analytical standard, ≥98% (TLC)
Supelco
Norfloxacin, VETRANAL®, analytical standard