Merck
Topography of transcriptionally active chromatin in glioblastoma.
Science advances (2021-05-02)
Liang Xu, Ye Chen, Yulun Huang, Edwin Sandanaraj, John S Yu, Ruby Yu-Tong Lin, Pushkar Dakle, Xin-Yu Ke, Yuk Kien Chong, Lynnette Koh, Anand Mayakonda, Kassoum Nacro, Jeffrey Hill, Mo-Li Huang, Sigal Gery, See Wee Lim, Zhengyun Huang, Ying Xu, Jianxiang Chen, Longchuan Bai, Shaomeng Wang, Hiroaki Wakimoto, Tseng Tsai Yeo, Beng Ti Ang, Markus Müschen, Carol Tang, Tuan Zea Tan, H Phillip Koeffler
ABSTRACT
Molecular profiling of the most aggressive brain tumor glioblastoma (GBM) on the basis of gene expression, DNA methylation, and genomic variations advances both cancer research and clinical diagnosis. The enhancer architectures and regulatory circuitries governing tumor-intrinsic transcriptional diversity and subtype identity are still elusive. Here, by mapping H3K27ac deposition, we analyze the active regulatory landscapes across 95 GBM biopsies, 12 normal brain tissues, and 38 cell line counterparts. Analyses of differentially regulated enhancers and super-enhancers uncovered previously unrecognized layers of intertumor heterogeneity. Integrative analysis of variant enhancer loci and transcriptome identified topographies of transcriptional enhancers and core regulatory circuitries in four molecular subtypes of primary tumors: AC1-mesenchymal, AC1-classical, AC2-proneural, and AC3-proneural. Moreover, this study reveals core oncogenic dependency on super-enhancer-driven transcriptional factors, long noncoding RNAs, and druggable targets in GBM. Through profiling of transcriptional enhancers, we provide clinically relevant insights into molecular classification, pathogenesis, and therapeutic intervention of GBM.
Sigma-Aldrich
Anti-MKNK2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-BCL6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-RFX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution