Merck
  • Cargo receptor-assisted endoplasmic reticulum export of pathogenic α1-antitrypsin polymers.
Cargo receptor-assisted endoplasmic reticulum export of pathogenic α1-antitrypsin polymers.
Cell reports (2021-05-20)
Adriana Ordóñez, Heather P Harding, Stefan J Marciniak, David Ron
ABSTRACT
Circulating polymers of α1-antitrypsin (α1AT) are neutrophil chemo-attractants and contribute to inflammation, yet cellular factors affecting their secretion remain obscure. We report on a genome-wide CRISPR-Cas9 screen for genes affecting trafficking of polymerogenic α1ATH334D. A CRISPR enrichment approach based on recovery of single guide RNA (sgRNA) sequences from phenotypically selected fixed cells reveals that cells with high-polymer content are enriched in sgRNAs targeting genes involved in "cargo loading into COPII-coated vesicles," where "COPII" is coat protein II, including the cargo receptors lectin mannose binding1 (LMAN1) and surfeit protein locus 4 (SURF4). LMAN1- and SURF4-disrupted cells display a secretion defect extending beyond α1AT monomers to polymers. Polymer secretion is especially dependent on SURF4 and correlates with a SURF4-α1ATH334D physical interaction and with their co-localization at the endoplasmic reticulum (ER). These findings indicate that ER cargo receptors co-ordinate progression of α1AT out of the ER and modulate the accumulation of polymeric α1AT not only by controlling the concentration of precursor monomers but also by promoting secretion of polymers.
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
FLI-06, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Supelco
N-(Propionyloxy)succinimide, for LC-MS derivatization, LiChropur, ≥95% (HPLC)