Merck
  • An Integrative Study of Protein-RNA Condensates Identifies Scaffolding RNAs and Reveals Players in Fragile X-Associated Tremor/Ataxia Syndrome.
An Integrative Study of Protein-RNA Condensates Identifies Scaffolding RNAs and Reveals Players in Fragile X-Associated Tremor/Ataxia Syndrome.
Cell reports (2018-12-20)
Fernando Cid-Samper, Mariona Gelabert-Baldrich, Benjamin Lang, Nieves Lorenzo-Gotor, Riccardo Delli Ponti, Lies-Anne W F M Severijnen, Benedetta Bolognesi, Ellen Gelpi, Renate K Hukema, Teresa Botta-Orfila, Gian Gaetano Tartaglia
ABSTRACT
Recent evidence indicates that specific RNAs promote the formation of ribonucleoprotein condensates by acting as scaffolds for RNA-binding proteins (RBPs). We systematically investigated RNA-RBP interaction networks to understand ribonucleoprotein assembly. We found that highly contacted RNAs are structured, have long UTRs, and contain nucleotide repeat expansions. Among the RNAs with such properties, we identified the FMR1 3' UTR that harbors CGG expansions implicated in fragile X-associated tremor/ataxia syndrome (FXTAS). We studied FMR1 binding partners in silico and in vitro and prioritized the splicing regulator TRA2A for further characterization. In a FXTAS cellular model, we validated the TRA2A-FMR1 interaction and investigated implications of its sequestration at both transcriptomic and post-transcriptomic levels. We found that TRA2A co-aggregates with FMR1 in a FXTAS mouse model and in post-mortem human samples. Our integrative study identifies key components of ribonucleoprotein aggregates, providing links to neurodegenerative disease and allowing the discovery of therapeutic targets.
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Millipore
Protein G, HRP conjugate, 500 µg Protein G, HRP conjugate for Dot Blot, ELISA, Western Blotting & IHC.
Sigma-Aldrich
Phosphate buffered saline, BioUltra, solution