Merck
  • Suppression of sphingosine 1-phosphate lyase retards the liver regeneration in mice after partial hepatectomy.
Suppression of sphingosine 1-phosphate lyase retards the liver regeneration in mice after partial hepatectomy.
Bioscience reports (2020-07-03)
Yuko Kageyama, Baasanjav Uranbileg, Yoshika Kusumoto, Eri Sakai, Hitoshi Ikeda, Makoto Kurano, Yutaka Yatomi
ABSTRACT
Liver regeneration is an extremely complicated process that is regulated by a number of signaling pathways. Sphingosine 1-phosphate (S1P), a potent bioactive lipid mediator playing crucial roles in various cellular responses through its receptors, has been attracting attention in the fields of hepatology, where S1P lyase (SPL), an irreversibly degrading enzyme of S1P, reportedly has a stimulatory role in growth of hepatocellular carcinoma (HCC). To examine whether SPL might play a stimulatory role in liver regeneration. Using in-vivo siRNA technology, we inhibited SPL expression. Seventy percent of the liver was resected in mice as partial hepatectomy (PH). Liver tissue samples were collected and mRNA expression level of the SPL, IHC of the proliferating cell nuclear antigen (PCNA), protein levels of various proliferation factors and lipid measurements were performed in different groups. The mRNA levels of SPL increased in PH mice on the third day after PH surgery. When we suppressed the expression of SPL by in-vivo siRNA, we observed a significant decline of the PCNA positive cell numbers. Furthermore, the Cyclin D1 expressions and phosphorylation of ERK also were decreased in the siSPL injected PH group. We verified the importance of the SPL in liver regeneration, using the mice PH model. SPL might be a potential target to facilitate liver regeneration.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-S1P lyase Antibody, from rabbit, purified by affinity chromatography