Merck
  • Injury triggers fascia fibroblast collective cell migration to drive scar formation through N-cadherin.
Injury triggers fascia fibroblast collective cell migration to drive scar formation through N-cadherin.
Nature communications (2020-11-08)
Dongsheng Jiang, Simon Christ, Donovan Correa-Gallegos, Pushkar Ramesh, Shruthi Kalgudde Gopal, Juliane Wannemacher, Christoph H Mayr, Valerio Lupperger, Qing Yu, Haifeng Ye, Martin Mück-Häusl, Vijayanand Rajendran, Li Wan, Juan Liu, Ursula Mirastschijski, Thomas Volz, Carsten Marr, Herbert B Schiller, Yuval Rinkevich
ABSTRACT
Scars are more severe when the subcutaneous fascia beneath the dermis is injured upon surgical or traumatic wounding. Here, we present a detailed analysis of fascia cell mobilisation by using deep tissue intravital live imaging of acute surgical wounds, fibroblast lineage-specific transgenic mice, and skin-fascia explants (scar-like tissue in a dish - SCAD). We observe that injury triggers a swarming-like collective cell migration of fascia fibroblasts that progressively contracts the skin and form scars. Swarming is exclusive to fascia fibroblasts, and requires the upregulation of N-cadherin. Both swarming and N-cadherin expression are absent from fibroblasts in the upper skin layers and the oral mucosa, tissues that repair wounds with minimal scar. Impeding N-cadherin binding inhibits swarming and skin contraction, and leads to reduced scarring in SCADs and in animals. Fibroblast swarming and N-cadherin thus provide therapeutic avenues to curtail fascia mobilisation and pathological fibrotic responses across a range of medical settings.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Benzyl ether, 98%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Trichrome Stain (Masson) Kit
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Bouin′s solution, histological fixative
Sigma-Aldrich
Anti-N-Cadherin antibody, Mouse monoclonal, clone GC-4, purified from hybridoma cell culture
Sigma-Aldrich
Weigert′s Iron Hematoxylin Set, Set
Sigma-Aldrich
Triton X-100, laboratory grade