Merck
  • IL4I1 Is a Metabolic Immune Checkpoint that Activates the AHR and Promotes Tumor Progression.
IL4I1 Is a Metabolic Immune Checkpoint that Activates the AHR and Promotes Tumor Progression.
Cell (2020-08-21)
Ahmed Sadik, Luis F Somarribas Patterson, Selcen Öztürk, Soumya R Mohapatra, Verena Panitz, Philipp F Secker, Pauline Pfänder, Stefanie Loth, Heba Salem, Mirja Tamara Prentzell, Bianca Berdel, Murat Iskar, Erik Faessler, Friederike Reuter, Isabelle Kirst, Verena Kalter, Kathrin I Foerster, Evelyn Jäger, Carina Ramallo Guevara, Mansour Sobeh, Thomas Hielscher, Gernot Poschet, Annekathrin Reinhardt, Jessica C Hassel, Marc Zapatka, Udo Hahn, Andreas von Deimling, Carsten Hopf, Rita Schlichting, Beate I Escher, Jürgen Burhenne, Walter E Haefeli, Naveed Ishaque, Alexander Böhme, Sascha Schäuble, Kathrin Thedieck, Saskia Trump, Martina Seiffert, Christiane A Opitz
ABSTRACT
Aryl hydrocarbon receptor (AHR) activation by tryptophan (Trp) catabolites enhances tumor malignancy and suppresses anti-tumor immunity. The context specificity of AHR target genes has so far impeded systematic investigation of AHR activity and its upstream enzymes across human cancers. A pan-tissue AHR signature, derived by natural language processing, revealed that across 32 tumor entities, interleukin-4-induced-1 (IL4I1) associates more frequently with AHR activity than IDO1 or TDO2, hitherto recognized as the main Trp-catabolic enzymes. IL4I1 activates the AHR through the generation of indole metabolites and kynurenic acid. It associates with reduced survival in glioma patients, promotes cancer cell motility, and suppresses adaptive immunity, thereby enhancing the progression of chronic lymphocytic leukemia (CLL) in mice. Immune checkpoint blockade (ICB) induces IDO1 and IL4I1. As IDO1 inhibitors do not block IL4I1, IL4I1 may explain the failure of clinical studies combining ICB with IDO1 inhibition. Taken together, IL4I1 blockade opens new avenues for cancer therapy.
Sigma-Aldrich
CRISPR/Cas9 Products and Services, Design and order CRISPR gRNA, Cas9, screening libraries, controls and companion products. Formats include plant, lentivirus, IVT-RNA, plasmid, synthetic, and protein.
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Kynurenic acid, ≥98%
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Bromophenol Blue sodium salt
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
Indole-3-pyruvic acid, ≥97%
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Coenzyme A sodium salt hydrate, cofactor for acyl transfer
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
Indole-3-carboxaldehyde, 97%
Sigma-Aldrich
O-(Carboxymethyl)hydroxylamine hemihydrochloride, 98%
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Ampliflu Red, for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Tryptophan-13C11,15N2, ≥99 atom %, ≥98% (CP)
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
DL-Indole-3-lactic acid, 99%
Sigma-Aldrich
4-Hydroxyphenylpyruvic acid, 98%
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)