Merck
  • Characteristics of a Hybrid Detector Combined with a Perovskite Active Layer for Indirect X-ray Detection.
Characteristics of a Hybrid Detector Combined with a Perovskite Active Layer for Indirect X-ray Detection.
Sensors (Basel, Switzerland) (2020-12-05)
Hailiang Liu, Jehoon Lee, Jungwon Kang
ABSTRACT
In this study, we investigated the characteristics of an organic-inorganic hybrid indirect-type X-ray detector with a CH3NH3PbI3 (MAPbI3) perovskite active layer. A layer with a thickness of 192 nm annealed at 100 °C showed higher absorption, higher crystallinity, and lower surface roughness than did perovskite layers made under different conditions. In the indirect X-ray detector, a scintillator coupled with the detector to convert X-ray photons to visible photons, and the converted photons were absorbed by the active layer to generate charge carriers. The detector with the optimized MAPbI3 (192 nm thick and 100 °C annealing condition) active layer was coupled with a CsI(Tl) scintillator which consisted of 400 μm thick CsI and 0.5 mm thick Al, and achieved the highest sensitivity, i.e., 2.84 mA/Gy·cm2. In addition, the highest short-circuit current density (JSC), i.e., 18.78 mA/cm2, and the highest mobility, i.e., 2.83 × 10-4 cm2/V·s, were obtained from the same detector without the CsI(Tl) scintillator.
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Lead(II) iodide, 99.999% trace metals basis
Sigma-Aldrich
Methylammonium iodide, 98%