Merck
  • Anti-Ro52 antibody acts on the S5-pore linker of hERG to chronically reduce channel expression.
Anti-Ro52 antibody acts on the S5-pore linker of hERG to chronically reduce channel expression.
Cardiovascular research (2018-12-14)
John Szendrey, Shawn M Lamothe, Stephanie Vanner, Jun Guo, Tonghua Yang, Wentao Li, Jordan Davis, Mala Joneja, Adrian Baranchuk, Shetuan Zhang
ABSTRACT
The human ether-a-go-go-related gene (hERG) encodes the rapidly activating delayed rectifier potassium channel (IKr). Malfunction of hERG/IKr is the primary cause of acquired long QT syndrome (LQTS), an electrical disorder of the heart that can cause arrhythmias and sudden death. Patients with autoimmune diseases display a high incidence of LQTS. While dysfunction of hERG channels induced by autoantibodies such as anti-Ro52 may play a role in this pathology, the underlying mechanisms are not well understood. Here, we investigated the acute and chronic effects of anti-Ro52 antibody on hERG channels stably expressed in human embryonic kidney (hERG-HEK) 293 cells as well as IKr in neonatal rat ventricular myocytes. Using whole-cell patch clamp, western blot analyses, and immunocytochemistry, we found that a 12-h treatment of hERG-HEK cells with patients' sera containing anti-Ro52 autoantibody decreased the hERG current (IhERG) by 32% compared to cells treated with autoantibody-negative patients' sera. Commercial anti-Ro52 antibody at 100 µg/mL did not acutely block IhERG. Instead, a 12-h treatment with anti-Ro52 antibody at a concentration of 4 µg/mL significantly reduced mature hERG protein expression and IhERG. Specifically, anti-Ro52 antibody did not acutely block hERG current but chronically facilitated hERG endocytic degradation. The extracellular S5-pore linker of hERG, which forms the turret of the channel on the outside of the cell, is the target region for anti-Ro52-mediated hERG reduction since its replacement with the analogous region of EAG abolished the anti-Ro52 effect. In neonatal rat ventricular myocytes, 100 µg/mL anti-Ro52 antibody did not acutely block IKr, but a 12-h treatment of cells with 4 µg/mL anti-Ro52 antibody selectively reduced IKr and prolonged the action potential duration. Our results indicate that anti-Ro52 antibody acts on the hERG S5-pore linker to chronically decrease hERG expression and current. These findings provide novel insights into hERG regulation and anti-Ro52 antibody-associated LQTS.
Sigma-Aldrich
Anti-TRIM21 (AB2) antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology