Merck
Androstene-3,5-dienes as ER-beta selective SERMs.
Bioorganic & medicinal chemistry letters (2007-09-25)
Timothy A Blizzard, Candido Gude, Jerry D Morgan, Wanda Chan, Elizabeth T Birzin, Marina Mojena, Consuelo Tudela, Fang Chen, Kristin Knecht, Qin Su, Bryan Kraker, Ralph T Mosley, Mark A Holmes, Susan P Rohrer, Milton L Hammond
ABSTRACT
A series of androstene-3,5-diene derivatives were prepared. Despite lacking the C-3 hydroxyl previously believed necessary for ER activity, some of the analogs retained surprising affinity for ER-beta. For example, diene 4 retained excellent selectivity and potency as an ER-beta agonist and was more selective for ER-beta over the androgen receptor (AR).
Product Description
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture