Merck
  • An autosomal dominant neurological disorder caused by de novo variants in FAR1 resulting in uncontrolled synthesis of ether lipids.
An autosomal dominant neurological disorder caused by de novo variants in FAR1 resulting in uncontrolled synthesis of ether lipids.
Genetics in medicine : official journal of the American College of Medical Genetics (2020-11-27)
Sacha Ferdinandusse, Kirsty McWalter, Heleen Te Brinke, Lodewijk IJlst, Petra M Mooijer, Jos P N Ruiter, Alida E M van Lint, Mia Pras-Raves, Eric Wever, Francisca Millan, Maria J Guillen Sacoto, Amber Begtrup, Mark Tarnopolsky, Lauren Brady, Roger L Ladda, Susan L Sell, Catherine B Nowak, Jessica Douglas, Cuixia Tian, Elizabeth Ulm, Seth Perlman, Arlene V Drack, Karen Chong, Nicole Martin, Jennifer Brault, Elly Brokamp, Camilo Toro, William A Gahl, Ellen F Macnamara, Lynne Wolfe, Quinten Waisfisz, Petra J G Zwijnenburg, Alban Ziegler, Magalie Barth, Rosemarie Smith, Sara Ellingwood, Deborah Gaebler-Spira, Somayeh Bakhtiari, Michael C Kruer, Antoine H C van Kampen, Ronald J A Wanders, Hans R Waterham, David Cassiman, Frédéric M Vaz
ABSTRACT
In this study we investigate the disease etiology in 12 patients with de novo variants in FAR1 all resulting in an amino acid change at position 480 (p.Arg480Cys/His/Leu). Following next-generation sequencing and clinical phenotyping, functional characterization was performed in patients' fibroblasts using FAR1 enzyme analysis, FAR1 immunoblotting/immunofluorescence, and lipidomics. All patients had spastic paraparesis and bilateral congenital/juvenile cataracts, in most combined with speech and gross motor developmental delay and truncal hypotonia. FAR1 deficiency caused by biallelic variants results in defective ether lipid synthesis and plasmalogen deficiency. In contrast, patients' fibroblasts with the de novo FAR1 variants showed elevated plasmalogen levels. Further functional studies in fibroblasts showed that these variants cause a disruption of the plasmalogen-dependent feedback regulation of FAR1 protein levels leading to uncontrolled ether lipid production. Heterozygous de novo variants affecting the Arg480 residue of FAR1 lead to an autosomal dominant disorder with a different disease mechanism than that of recessive FAR1 deficiency and a diametrically opposed biochemical phenotype. Our findings show that for patients with spastic paraparesis and bilateral cataracts, FAR1 should be considered as a candidate gene and added to gene panels for hereditary spastic paraplegia, cerebral palsy, and juvenile cataracts.
Sigma-Aldrich
Anti-PMP70 antibody, Mouse monoclonal, clone 70-18, purified from hybridoma cell culture
Sigma-Aldrich
Anti-FAR1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid