Merck
  • Metal complexes with superoxide dismutase-like activity as candidates for anti-prion drug.
Metal complexes with superoxide dismutase-like activity as candidates for anti-prion drug.
Bioorganic & medicinal chemistry letters (2006-09-22)
Tomoko Fukuuchi, Katsumi Doh-Ura, Shin'ichi Yoshihara, Shigeru Ohta
ABSTRACT
Various compounds were evaluated for ability to inhibit the formation of the abnormal protease-resistant form of prion protein (PrP-res) in two cell lines infected with different prion strains. Examination of the structure-activity relationships indicated that compounds with copper-selective chelating ability and whose copper complexes have high SOD-like activity are candidates for anti-prion drug.
Sigma-Aldrich
8-Quinolinol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
Neocuproine, ≥98%
Supelco
8-Quinolinol, PESTANAL®, analytical standard
Sigma-Aldrich
8-Hydroxyquinoline, ACS reagent, 98.5%
Sigma-Aldrich
Bathocuproine, 96%
Sigma-Aldrich
8-Hydroxyquinoline