Merck
  • Nonthermal plasma treatment of polymers modulates biological fouling but can cause material embrittlement.
Nonthermal plasma treatment of polymers modulates biological fouling but can cause material embrittlement.
Journal of the mechanical behavior of biomedical materials (2020-10-13)
Greg D Learn, Emerson J Lai, Emily J Wilson, Horst A von Recum
ABSTRACT
Plasma-based treatment is a prevalent strategy to alter biological response and enhance biomaterial coating quality at the surfaces of biomedical devices and implants, especially polymeric materials. Plasma, an ionized gas, is often thought to have negligible effects on the bulk properties of prosthetic substrates given that it alters the surface chemistry on only the outermost few nanometers of material. However, no studies to date have systematically explored the effects of plasma exposure on both the surface and bulk properties of a biomaterial. This work examines the time-dependent effects of a nonthermal plasma on the surface and bulk (i.e. mechanical) properties of polymeric implants, specifically polypropylene surgical meshes and sutures. Findings suggest that plasma exposure improved resistance to fibrinogen adsorption and Escherichia coli attachment, and promoted mammalian fibroblast attachment, although increased duration of exposure resulted in a state of diminishing returns. At the same time, it was observed that plasma exposure can be detrimental to the material properties of individual filaments (i.e. sutures), as well as the structural characteristics of knitted meshes, with longer exposures resulting in further embrittlement and larger changes in anisotropic behavior. Though there are few guidelines regarding appropriate mechanical properties of surgical textiles, the results from this investigation imply that there are ultimate exposure limits for plasma-based treatments of polymeric implant materials when structural properties must be preserved, and that the effects of a plasma on a given biomaterial should be examined carefully before translation to a clinical scenario.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)