Merck
  • Novel 6-O-acylated vitamin C derivatives as hyaluronidase inhibitors with selectivity for bacterial lyases.
Novel 6-O-acylated vitamin C derivatives as hyaluronidase inhibitors with selectivity for bacterial lyases.
Bioorganic & medicinal chemistry letters (2006-08-16)
Martin Spickenreither, Stephan Braun, Günther Bernhardt, Stefan Dove, Armin Buschauer
ABSTRACT
Previously, we identified ascorbic acid 6-O-hexadecanoate as an up to 1500 times more potent inhibitor of bacterial and bovine hyaluronidases than the parent compound, vitamin C, and determined a crystal structure of hyaluronidase from Streptococcus pneumoniae in complex with the inhibitor. As the alkanoyl chain interacts with a hydrophobic patch of the enzyme we synthesized other 6-O-acylated vitamin C derivatives bearing various lipophilic residues and investigated the inhibition of Streptococcus agalactiae strain 4755 hyaluronidase (SagHyal(4755)) and of bovine testicular hyaluronidases (BTH) in a turbidimetric assay. All compounds showed selectivity for the bacterial enzyme. Whereas vitamin C 6-O-hexanoate only weakly inhibited SagHyal(4755), the inhibition of both enzymes increased with the length of the aliphatic chain. In the case of the 6-O-octadecanoate, IC(50) values of 0.9 and 39microM for SagHyal(4755) and BTH, respectively, were determined. Partial replacement of the aliphatic chain with a phenyl, p-phenylene or p-biphenylyl group resulted in inhibitors with activity in the lower micromolar range, too. The title compounds are among the most potent inhibitors of both enzymes known to date.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Ascorbic acid 6-palmitate, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)