Merck
  • Parallel artificial membrane permeability assay: a new membrane for the fast prediction of passive human skin permeability.
Parallel artificial membrane permeability assay: a new membrane for the fast prediction of passive human skin permeability.
Journal of medicinal chemistry (2006-06-23)
Giorgio Ottaviani, Sophie Martel, Pierre-Alain Carrupt
ABSTRACT
This work was devoted to the search for new artificial membranes allowing a rapid evaluation of passive human skin permeation of compounds with a parallel artificial membrane permeability assay (PAMPA). Effective permeability coefficients (Pe) determined for a set of compounds using the PAMPA technique with isopropyl myristate (IPM) and silicone oil, alone or in mixture, were compared to the corresponding human skin permeability coefficient values (Kp). A good correlation between Pe and Kp was found for compounds tested through a membrane consisting of 70% silicone and 30% IPM. Moreover, positive correlation between the membrane retention of compounds and stratum corneum/water partition coefficients (PSC) was established. These results showed that this new artificial membrane, defined as PAMPA-skin, is able to mimic the main barrier properties of human stratum corneum and can be used for the fast prediction of passive human skin permeability coefficients.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
4-Ethylphenol, 99%
Sigma-Aldrich
Methyl nicotinate, 99%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Methyl nicotinate, puriss., ≥99.0% (GC)
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
Resorcinol, ACS reagent, ≥99.0%
Sigma-Aldrich
4-Chlorophenol, ≥99%
Sigma-Aldrich
Isoquinoline, 97%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
4-Ethylphenol, ≥97.0% (GC)
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Progesterone, ≥99%
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture