- Serotonin and vasotocin function in territoriality.
Serotonin and vasotocin function in territoriality.
This ethopharmacological investigation comprised a long-term field study that examined the function of serotonergic and vasotonergic systems in territoriality. Adult territorial and non-territorial (silent) male coquí frogs (Eleutherodactylus coqui) were injected (IP) with either arginine vasotocin (AVT) or one of two serotonin agonists, 5-HT2A/2C selective agonist, (±) DOI - [(±)-1-(2,5-dimethoxy-4-iodophenyl)-2-aminopropane], or 2) the 5-HT1A selective agonist, 8-OH-DPAT - [(±)-2-dipropylamino-8-hydroxy-1,2,3,4-tetrahydronaphthalene]. Control groups received saline injections. Each male received two injections. Following the first injection, whether AVT or a 5HT agonist, the male was observed so that behavior could be documented prior to the second injection, which consisted of the other drug class. All frogs were marked, placed back in the exact location as captured, and observed for all behaviors and vocalizations. Territoriality in E. coqui includes several behavioral components: movement into a calling site, presentation of dominant physical displays, emitting advertisement calls, and defense a territory (including the use of physical force and/or aggressive vocalizations). This investigation found that particular territorial behaviors were significantly influenced by 5HT and AVT action. Initiation of advertisement calling is activated by AVT and suppressed by 5HT, calling rate is affected by 5HT activation, presentation of dominant physical displays are activated by AVT and repressed by 5HT activation, and movement associated with activation of territorial behavior is stimulated by AVT. These data suggested that both 5HT and AVT have a profound impact on territoriality and are two fundamental neuroendocrine systems that govern territorial behavior in social systems.