Merck
Objective assessment of stored blood quality by deep learning.
Proceedings of the National Academy of Sciences of the United States of America (2020-08-26)
Minh Doan, Joseph A Sebastian, Juan C Caicedo, Stefanie Siegert, Aline Roch, Tracey R Turner, Olga Mykhailova, Ruben N Pinto, Claire McQuin, Allen Goodman, Michael J Parsons, Olaf Wolkenhauer, Holger Hennig, Shantanu Singh, Anne Wilson, Jason P Acker, Paul Rees, Michael C Kolios, Anne E Carpenter
ABSTRACT
Stored red blood cells (RBCs) are needed for life-saving blood transfusions, but they undergo continuous degradation. RBC storage lesions are often assessed by microscopic examination or biochemical and biophysical assays, which are complex, time-consuming, and destructive to fragile cells. Here we demonstrate the use of label-free imaging flow cytometry and deep learning to characterize RBC lesions. Using brightfield images, a trained neural network achieved 76.7% agreement with experts in classifying seven clinically relevant RBC morphologies associated with storage lesions, comparable to 82.5% agreement between different experts. Given that human observation and classification may not optimally discern RBC quality, we went further and eliminated subjective human annotation in the training step by training a weakly supervised neural network using only storage duration times. The feature space extracted by this network revealed a chronological progression of morphological changes that better predicted blood quality, as measured by physiological hemolytic assay readouts, than the conventional expert-assessed morphology classification system. With further training and clinical testing across multiple sites, protocols, and instruments, deep learning and label-free imaging flow cytometry might be used to routinely and objectively assess RBC storage lesions. This would automate a complex protocol, minimize laboratory sample handling and preparation, and reduce the impact of procedural errors and discrepancies between facilities and blood donors. The chronology-based machine-learning approach may also improve upon humans' assessment of morphological changes in other biomedically important progressions, such as differentiation and metastasis.
Sigma-Aldrich
Clear-view Snap-Cap microtubes, capacity 1.5 mL, natural, low retention