Merck
  • Bioaccessibility and cellular uptake by Caco-2 cells of carotenoids and chlorophylls from orange peels: A comparison between conventional and ionic liquid mediated extractions.
Bioaccessibility and cellular uptake by Caco-2 cells of carotenoids and chlorophylls from orange peels: A comparison between conventional and ionic liquid mediated extractions.
Food chemistry (2020-08-28)
Daniella C Murador, Leonardo M De Souza Mesquita, Bruna V Neves, Anna R C Braga, Paula L G Martins, Leila Q Zepka, Veridiana V De Rosso
ABSTRACT
Native extracts from orange peels were obtained by a conventional method using acetone and, an alternative method using ionic liquid (1-butyl-3-methylimidazolium chloride ([C4mim]Cl)). The bioaccessibilities and cellular uptakes of carotenoids, esters and chlorophylls were evaluated, since the influence of esterification on bioaccessibility and bioavailability is not well established. For this, the extracts were emulsified, submitted to in vitro simulated digestion model according to the INFOGEST protocol, followed by uptake by Caco-2 cells. Compounds were separated, identified and quantified by HPLC-PDA-MS/MS. After digestion, 22.0% and 26.2% of the total carotenoids and 45.9% and 68.7% of the chlorophylls were bioaccessible from the acetone and [C4mim]Cl extracts, respectively. The bioaccessibilities of xanthophylls and carotenes were significantly higher than those of the mono- and diesters. The uptake by Caco-2 cells varied from 130.2 to 131.9 ng/mg cell protein for total carotenoids and from 243.8 to 234.2 ng/mg cell protein for chlorophylls in the acetone and [C4mim]Cl extracts, respectively. In general, xanthophylls and esters were better absorbed than carotenes.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Supelco
Amberlite XAD7HP, 20-60 mesh
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
Sigma-Aldrich
MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
Fetal Bovine Serum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol Esterase from porcine pancreas, lyophilized, powder, white, ~35 U/mg