Merck
  • HDAC inhibitors elicit metabolic reprogramming by targeting super-enhancers in glioblastoma models.
HDAC inhibitors elicit metabolic reprogramming by targeting super-enhancers in glioblastoma models.
The Journal of clinical investigation (2020-04-22)
Trang Thi Thu Nguyen, Yiru Zhang, Enyuan Shang, Chang Shu, Consuelo Torrini, Junfei Zhao, Elena Bianchetti, Angeliki Mela, Nelson Humala, Aayushi Mahajan, Arif O Harmanci, Zhengdeng Lei, Mark Maienschein-Cline, Catarina M Quinzii, Mike-Andrew Westhoff, Georg Karpel-Massler, Jeffrey N Bruce, Peter Canoll, Markus D Siegelin
ABSTRACT
The Warburg effect is a tumor-related phenomenon that could potentially be targeted therapeutically. Here, we showed that glioblastoma (GBM) cultures and patients' tumors harbored super-enhancers in several genes related to the Warburg effect. By conducting a transcriptome analysis followed by ChIP-Seq coupled with a comprehensive metabolite analysis in GBM models, we found that FDA-approved global (panobinostat, vorinostat) and selective (romidepsin) histone deacetylase (HDAC) inhibitors elicited metabolic reprogramming in concert with disruption of several Warburg effect-related super-enhancers. Extracellular flux and carbon-tracing analyses revealed that HDAC inhibitors blunted glycolysis in a c-Myc-dependent manner and lowered ATP levels. This resulted in the engagement of oxidative phosphorylation (OXPHOS) driven by elevated fatty acid oxidation (FAO), rendering GBM cells dependent on these pathways. Mechanistically, interference with HDAC1/-2 elicited a suppression of c-Myc protein levels and a concomitant increase in 2 transcriptional drivers of oxidative metabolism, PGC1α and PPARD, suggesting an inverse relationship. Rescue and ChIP experiments indicated that c-Myc bound to the promoter regions of PGC1α and PPARD to counteract their upregulation driven by HDAC1/-2 inhibition. Finally, we demonstrated that combination treatment with HDAC and FAO inhibitors extended animal survival in patient-derived xenograft model systems in vivo more potently than single treatments in the absence of toxicity.
Sigma-Aldrich
MISSION® esiRNA, targeting human CPT1A
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC2
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC1
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
MISSION® esiRNA, targeting human PPARD