- Fillers as Heaters for Photothermal Polymerization upon NIR Light.
Fillers as Heaters for Photothermal Polymerization upon NIR Light.
Photo-induced thermal polymerization upon near-infrared (NIR) light irradiation has been reported in the literature. In this approach, a component able to convert the NIR light into heat must be used in combination with a thermal initiator to initiate the free-radical polymerization of (meth)acrylates. In recent studies, some absorbers have been presented as very efficient heat generators (called heaters). In the present work, different fillers are investigated as heaters and compared to organic NIR absorbers. An alkoxyamine (e.g., BlocBuilder-MA) is used as thermal initiator and is dissociated by the heat generated by the NIR photoexcitation of the fillers. In the present work, several fillers are examined: graphene oxide, graphene nanoplatelets, multi-walled carbon nanotubes, and silicon carbide. Due to the energy of the photon delivered, NIR light curing is challenging but offers several advantages compared to visible light. The most interesting feature is the deeper penetration of the light inside the photocurable resin, enabling the polymerization of thick samples. Parallel to this, incorporation of fillers in resins allows unique access to composites through photothermal polymerization of (meth)acrylates. Three different wavelengths of irradiation have been studied: 785, 940, and 1064 nm.