Merck
  • CN133, a Novel Brain-Penetrating Histone Deacetylase Inhibitor, Hampers Tumor Growth in Patient-Derived Pediatric Posterior Fossa Ependymoma Models.
CN133, a Novel Brain-Penetrating Histone Deacetylase Inhibitor, Hampers Tumor Growth in Patient-Derived Pediatric Posterior Fossa Ependymoma Models.
Cancers (2020-07-28)
Roberta Antonelli, Carlos Jiménez, Misha Riley, Tiziana Servidei, Riccardo Riccardi, Aroa Soriano, Josep Roma, Elena Martínez-Saez, Maurizio Martini, Antonio Ruggiero, Lucas Moreno, Josep Sánchez de Toledo, Soledad Gallego, Jordi Bové, Jacob M Hooker, Miguel F Segura
ABSTRACT
Pediatric ependymoma (EPN) is a highly aggressive tumor of the central nervous system that remains incurable in 40% of cases. In children, the majority of cases develop in the posterior fossa and can be classified into two distinct molecular entities: EPN posterior fossa A (PF-EPN-A) and EPN posterior fossa B (PF-EPN-B). Patients with PF-EPN-A have poor outcome and are in demand of new therapies. In general, PF-EPN-A tumors show a balanced chromosome copy number profile and have no recurrent somatic nucleotide variants. However, these tumors present abundant epigenetic deregulations, thereby suggesting that epigenetic therapies could provide new opportunities for PF-EPN-A patients. In vitro epigenetic drug screening of 11 compounds showed that histone deacetylase inhibitors (HDACi) had the highest anti-proliferative activity in two PF-EPN-A patient-derived cell lines. Further screening of 5 new brain-penetrating HDACi showed that CN133 induced apoptosis in vitro, reduced tumor growth in vivo and significantly extended the survival of mice with orthotopically-implanted EPN tumors by modulation of the unfolded protein response, PI3K/Akt/mTOR signaling, and apoptotic pathways among others. In summary, our results provide solid preclinical evidence for the use of CN133 as a new therapeutic agent against PF-EPN-A tumors.
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
UNC1999, ≥98% (HPLC)
Sigma-Aldrich
UNC0642, ≥98% (HPLC)