Merck
  • LUF7244 plus Dofetilide Rescues Aberrant Kv11.1 Trafficking and Produces Functional IKv11.1.
LUF7244 plus Dofetilide Rescues Aberrant Kv11.1 Trafficking and Produces Functional IKv11.1.
Molecular pharmacology (2020-04-04)
Muge Qile, Yuan Ji, Tyona D Golden, Marien J C Houtman, Fee Romunde, Doreth Fransen, Willem B van Ham, Ad P IJzerman, Craig T January, Laura H Heitman, Anna Stary-Weinzinger, Brian P Delisle, Marcel A G van der Heyden
ABSTRACT
Voltage-gated potassium 11.1 (Kv11.1) channels play a critical role in repolarization of cardiomyocytes during the cardiac action potential (AP). Drug-mediated Kv11.1 blockade results in AP prolongation, which poses an increased risk of sudden cardiac death. Many drugs, like pentamidine, interfere with normal Kv11.1 forward trafficking and thus reduce functional Kv11.1 channel densities. Although class III antiarrhythmics, e.g., dofetilide, rescue congenital and acquired forward trafficking defects, this is of little use because of their simultaneous acute channel blocking effect. We aimed to test the ability of a combination of dofetilide plus LUF7244, a Kv11.1 allosteric modulator/activator, to rescue Kv11.1 trafficking and produce functional Kv11.1 current. LUF7244 treatment by itself did not disturb or rescue wild type (WT) or G601S-Kv11.1 trafficking, as shown by Western blot and immunofluorescence microcopy analysis. Pentamidine-decreased maturation of WT Kv11.1 levels was rescued by 10 μM dofetilide or 10 μM dofetilide + 5 μM LUF7244. In trafficking defective G601S-Kv11.1 cells, dofetilide (10 μM) or dofetilide + LUF7244 (10 + 5 μM) also restored Kv11.1 trafficking, as demonstrated by Western blot and immunofluorescence microscopy. LUF7244 (10 μM) increased IKv 11.1 despite the presence of dofetilide (1 μM) in WT Kv11.1 cells. In G601S-expressing cells, long-term treatment (24-48 hour) with LUF7244 (10 μM) and dofetilide (1 μM) increased IKv11.1 compared with nontreated or acutely treated cells. We conclude that dofetilide plus LUF7244 rescues Kv11.1 trafficking and produces functional IKv11.1 Thus, combined administration of LUF7244 and an IKv11.1 trafficking corrector could serve as a new pharmacological therapy of both congenital and drug-induced Kv11.1 trafficking defects. SIGNIFICANCE STATEMENT: Decreased levels of functional Kv11.1 potassium channel at the plasma membrane of cardiomyocytes prolongs action potential repolarization, which associates with cardiac arrhythmia. Defective forward trafficking of Kv11.1 channel protein is an important factor in acquired and congenital long QT syndrome. LUF7244 as a negative allosteric modulator/activator in combination with dofetilide corrected both congenital and acquired Kv11.1 trafficking defects, resulting in functional Kv11.1 current.
Sigma-Aldrich
LUF7244, ≥98% (HPLC)
Pricing and availability is not currently available.