Merck
  • Foot-and-mouth disease virus can induce a specific and rapid CD4+ T-cell-independent neutralizing and isotype class-switched antibody response in naïve cattle.
Foot-and-mouth disease virus can induce a specific and rapid CD4+ T-cell-independent neutralizing and isotype class-switched antibody response in naïve cattle.
Journal of virology (2009-01-30)
Nicholas Juleff, Miriam Windsor, Eric A Lefevre, Simon Gubbins, Pip Hamblin, Elizabeth Reid, Kerry McLaughlin, Peter C L Beverley, Ivan W Morrison, Bryan Charleston
ABSTRACT
The role of T-lymphocyte subsets in recovery from foot-and-mouth disease virus (FMDV) infection in calves was investigated by administering subset-specific monoclonal antibodies. The depletion of circulating CD4(+) or WC1(+) gammadelta T cells was achieved for a period extending from before challenge to after resolution of viremia and peak clinical signs, whereas CD8(+) cell depletion was only partial. The depletion of CD4(+) cells was also confirmed by analysis of lymph node biopsy specimens 5 days postchallenge. Depletion with anti-WC1 and anti-CD8 antibodies had no effect on the kinetics of infection, clinical signs, and immune responses following FMDV infection. Three of the four CD4(+) T-cell-depleted calves failed to generate an antibody response to the nonstructural polyprotein 3ABC but generated a neutralizing antibody response similar to that in the controls, including rapid isotype switching to immunoglobulin G antibody. We conclude that antibody responses to sites on the surface of the virus capsid are T cell independent, whereas those directed against the nonstructural proteins are T cell dependent. CD4 depletion was found to substantially inhibit antibody responses to the G-H peptide loop VP1(135-156) on the viral capsid, indicating that responses to this particular site, which has a more mobile structure than other neutralizing sites on the virus capsid, are T cell dependent. The depletion of CD4(+) T cells had no adverse effect on the magnitude or duration of clinical signs or clearance of virus from the circulation. Overall, we conclude that CD4(+) T-cell-independent antibody responses play a major role in the resolution of foot-and-mouth disease in cattle.
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 1 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 4 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, peroxidase substrate
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 3 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 15 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077 g/mL
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 60 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 2 mg substrate per tablet