Merck
Alpha-synuclein and tau: teammates in neurodegeneration?
Molecular neurodegeneration (2014-10-30)
Simon Moussaud, Daryl R Jones, Elisabeth L Moussaud-Lamodière, Marion Delenclos, Owen A Ross, Pamela J McLean
ABSTRACT
The accumulation of α-synuclein aggregates is the hallmark of Parkinson's disease, and more generally of synucleinopathies. The accumulation of tau aggregates however is classically found in the brains of patients with dementia, and this type of neuropathological feature specifically defines the tauopathies. Nevertheless, in numerous cases α-synuclein positive inclusions are also described in tauopathies and vice versa, suggesting a co-existence or crosstalk of these proteinopathies. Interestingly, α-synuclein and tau share striking common characteristics suggesting that they may work in concord. Tau and α-synuclein are both partially unfolded proteins that can form toxic oligomers and abnormal intracellular aggregates under pathological conditions. Furthermore, mutations in either are responsible for severe dominant familial neurodegeneration. Moreover, tau and α-synuclein appear to promote the fibrillization and solubility of each other in vitro and in vivo. This suggests that interactions between tau and α-synuclein form a deleterious feed-forward loop essential for the development and spreading of neurodegeneration. Here, we review the recent literature with respect to elucidating the possible links between α-synuclein and tau.
Sigma-Aldrich
Tau-412 human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE), lyophilized powder