Merck
  • High-Throughput Screens of PAM-Flexible Cas9 Variants for Gene Knockout and Transcriptional Modulation.
High-Throughput Screens of PAM-Flexible Cas9 Variants for Gene Knockout and Transcriptional Modulation.
Cell reports (2020-03-05)
Mateusz Legut, Zharko Daniloski, Xinhe Xue, Dayna McKenzie, Xinyi Guo, Hans-Hermann Wessels, Neville E Sanjana
ABSTRACT
A key limitation of the widely used CRISPR enzyme S. pyogenes Cas9 is the strict requirement of an NGG protospacer-adjacent motif (PAM) at the target site. This constraint can be limiting for genome editing applications that require precise Cas9 positioning. Recently, two Cas9 variants with a relaxed PAM requirement (NG) have been developed (xCas9 and Cas9-NG), but their activity has been measured at only a small number of endogenous sites. Here, we devise a high-throughput Cas9 pooled competition screen to compare the performance of Cas9 variants at thousands of genomic loci for gene knockout, transcriptional activation, and inhibition. We show that PAM flexibility comes at a substantial cost of decreased DNA targeting and cleavage. Of the PAM-flexible variants, we find that Cas9-NG outperforms xCas9 regardless of genome engineering modality or PAM. Finally, we combine xCas9 mutations with those of Cas9-NG, creating a stronger transcriptional modulator than existing PAM-flexible Cas9 variants.
Sigma-Aldrich
Anti-2A peptide Antibody, clone 3H4, clone 3H4, from mouse
Pricing and availability is not currently available.
Millipore
Millipore® Steriflip® Vacuum Tube Top Filter, pore size 0.45 μm, PVDF membrane, funnel capacity 50 mL, pack of 25 ea
Pricing and availability is not currently available.