Merck
  • Exploring Antioxidant and Enzymes (A-Amylase and B-Glucosidase) Inhibitory Activity of Morinda lucida and Momordica charantia Leaves from Benin.
Exploring Antioxidant and Enzymes (A-Amylase and B-Glucosidase) Inhibitory Activity of Morinda lucida and Momordica charantia Leaves from Benin.
Foods (Basel, Switzerland) (2020-04-09)
Michaelle Chokki, Mihaela Cudălbeanu, Cheikna Zongo, Durand Dah-Nouvlessounon, Ioana Otilia Ghinea, Bianca Furdui, Robert Raclea, Aly Savadogo, Lamine Baba-Moussa, Sorin Marius Avamescu, Rodica Mihaela Dinica, Farid Baba-Moussa
ABSTRACT
Momordica charantia Linn. (Cucurbitaceae), the wild variety of bitter melon and Morinda lucida Benth (Rubiaceae) were commonly used as a popular folk medicine in Benin. This research focused to measure the antioxidant and enzyme inhibitory effects of M. charantia and M. lucida leaves and their antidiabetic activity. Antioxidant activities were evaluated by micro-dilution technique using DPPH free radical scavenging activity and β-carotene-linoleate bleaching assay. The α-amylase inhibition assay was carried out utilizing the 3,5-dinitrosalicylic acid procedure, while β-glucosidase inhibition assay was demonstrated using as substrate p-nitrophenyl-β-D-glucopyranoside (PNPG). HPLC-DAD analysis was realized using a high-performance liquid chromatography systems with diode-array detector, L-3000. Chlorogenic acid, epicatechin, daidzein, rutin, naringin, quercetin, naringenin and genistein were identified as polyphenol compounds in the both plants extract. Dichloromethane and ethyl acetate extracts showed a good α-amylase inhibitory activity (56.46 ± 1.96% and 58.76 ± 2.74% respectively). M. lucida methanolic extract has shown IC50 of 0.51 ± 0.01 mg/mL, which is the lowest for DPPH scavenging activity. M. lucida dichloromethane extract showed the highest inhibitory capacity of β-glucosidase activity (82.11. ± 2.15%). These results justify some traditional medicinal uses of both plants. The purified fractions could be used in future formulations, possibly incorporated in functional foods to combat certain diseases.
Sigma-Aldrich
Acarbose, ≥95% (HPLC)
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
Sigma-Aldrich
β-Glucosidase from almonds, lyophilized powder, ≥2 units/mg solid