Merck
  • Comparative Mortality and Adaptation of a Smurf Assay in two Species of Tenebrionid Beetles Exposed to Bacillus thuringiensis.
Comparative Mortality and Adaptation of a Smurf Assay in two Species of Tenebrionid Beetles Exposed to Bacillus thuringiensis.
Insects (2020-04-30)
Caroline Zanchi, Ana Sofia Lindeza, Joachim Kurtz
ABSTRACT
Bacillus thuringiensis is a spore-forming bacterium which infects insect larvae naturally via the oral route. Its virulence factors interact with the epithelium of the digestive tract of insect larvae, disrupting its function and eventually leading to the death of susceptible hosts. The most cited B. thuringiensis killing mechanism is the extensive damage caused to the insect midgut, leading to its leakage. The mortality caused by B. thuringiensis has been shown to vary between serovars and isolates, as well as between host life stages. Moreover, whether susceptibility to B. thuringiensis-induced gut leakage is generalized to all host species and whether there is individual variation within species is unclear. In this study, we adapted a non-invasive "Smurf" assay from Drosophila melanogaster to two species of tenebrionid beetles: The mealworm beetle Tenebriomolitor and the red flour beetle Triboliumcastaneum, during exposure to B. thuringiensis. We highlight a differential mortality between two age/size classes of T.molitor larvae, as well as different killing dynamics between B. thuringiensis var. tenebrionis and var. tolworthi in T.castaneum. The Smurf assay did not reveal a high occurrence of extensive gut disintegration in both host species upon ingestion during B. thuringiensis exposure.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Calcofluor White Stain, suitable for microbiology
Sigma-Aldrich
Erioglaucine disodium salt, coloring dye
Sigma-Aldrich
Albumin from chicken egg white, lyophilized powder, ≥98% (agarose gel electrophoresis)