Merck
  • BCG Vaccination in Humans Elicits Trained Immunity via the Hematopoietic Progenitor Compartment.
BCG Vaccination in Humans Elicits Trained Immunity via the Hematopoietic Progenitor Compartment.
Cell host & microbe (2020-06-17)
Branko Cirovic, L Charlotte J de Bree, Laszlo Groh, Bas A Blok, Joyce Chan, Walter J F M van der Velden, M E J Bremmers, Reinout van Crevel, Kristian Händler, Simone Picelli, Jonas Schulte-Schrepping, Kathrin Klee, Marije Oosting, Valerie A C M Koeken, Jakko van Ingen, Yang Li, Christine S Benn, Joachim L Schultze, Leo A B Joosten, Nigel Curtis, Mihai G Netea, Andreas Schlitzer
ABSTRACT
Induction of trained immunity by Bacille-Calmette-Guérin (BCG) vaccination mediates beneficial heterologous effects, but the mechanisms underlying its persistence and magnitude remain elusive. In this study, we show that BCG vaccination in healthy human volunteers induces a persistent transcriptional program connected to myeloid cell development and function within the hematopoietic stem and progenitor cell (HSPC) compartment in the bone marrow. We identify hepatic nuclear factor (HNF) family members 1a and b as crucial regulators of this transcriptional shift. These findings are corroborated by higher granulocyte numbers in BCG-vaccinated infants, HNF1 SNP variants that correlate with trained immunity, and elevated serum concentrations of the HNF1 target alpha-1 antitrypsin. Additionally, transcriptomic HSPC remodeling was epigenetically conveyed to peripheral CD14+ monocytes, displaying an activated transcriptional signature three months after BCG vaccination. Taken together, transcriptomic, epigenomic, and functional reprogramming of HSPCs and peripheral monocytes is a hallmark of BCG-induced trained immunity in humans.
Sigma-Aldrich
Human Serpin A1 ELISA Kit, for cell culture supernatants, plasma, and serum samples
Sigma-Aldrich
Rat serum
Sigma-Aldrich
Mouse serum