Merck
  • LncRNA-dependent nuclear stress bodies promote intron retention through SR protein phosphorylation.
LncRNA-dependent nuclear stress bodies promote intron retention through SR protein phosphorylation.
The EMBO journal (2019-11-30)
Kensuke Ninomiya, Shungo Adachi, Tohru Natsume, Junichi Iwakiri, Goro Terai, Kiyoshi Asai, Tetsuro Hirose
ABSTRACT
A number of long noncoding RNAs (lncRNAs) are induced in response to specific stresses to construct membrane-less nuclear bodies; however, their function remains poorly understood. Here, we report the role of nuclear stress bodies (nSBs) formed on highly repetitive satellite III (HSATIII) lncRNAs derived from primate-specific satellite III repeats upon thermal stress exposure. A transcriptomic analysis revealed that depletion of HSATIII lncRNAs, resulting in elimination of nSBs, promoted splicing of 533 retained introns during thermal stress recovery. A HSATIII-Comprehensive identification of RNA-binding proteins by mass spectrometry (ChIRP-MS) analysis identified multiple splicing factors in nSBs, including serine and arginine-rich pre-mRNA splicing factors (SRSFs), the phosphorylation states of which affect splicing patterns. SRSFs are rapidly de-phosphorylated upon thermal stress exposure. During stress recovery, CDC like kinase 1 (CLK1) was recruited to nSBs and accelerated the re-phosphorylation of SRSF9, thereby promoting target intron retention. Our findings suggest that HSATIII-dependent nSBs serve as a conditional platform for phosphorylation of SRSFs by CLK1 to promote the rapid adaptation of gene expression through intron retention following thermal stress exposure.
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%