Merck
  • Arhgef5 Binds α-Dystrobrevin 1 and Regulates Neuromuscular Junction Integrity.
Arhgef5 Binds α-Dystrobrevin 1 and Regulates Neuromuscular Junction Integrity.
Frontiers in molecular neuroscience (2020-06-27)
Krzysztof M Bernadzki, Patrycja Daszczuk, Katarzyna O Rojek, Marcin Pęziński, Marta Gawor, Bhola S Pradhan, Teresa de Cicco, Monika Bijata, Krystian Bijata, Jakub Włodarczyk, Tomasz J Prószyński, Paweł Niewiadomski
ABSTRACT
The neuromuscular junctions (NMJs) connect muscle fibers with motor neurons and enable the coordinated contraction of skeletal muscles. The dystrophin-associated glycoprotein complex (DGC) is an essential component of the postsynaptic machinery of the NMJ and is important for the maintenance of NMJ structural integrity. To identify novel proteins that are important for NMJ organization, we performed a mass spectrometry-based screen for interactors of α-dystrobrevin 1 (aDB1), one of the components of the DGC. The guanidine nucleotide exchange factor (GEF) Arhgef5 was found to be one of the aDB1 binding partners that is recruited to Tyr-713 in a phospho-dependent manner. We show here that Arhgef5 localizes to the NMJ and that its genetic depletion in the muscle causes the fragmentation of the synapses in conditional knockout mice. Arhgef5 loss in vivo is associated with a reduction in the levels of active GTP-bound RhoA and Cdc42 GTPases, highlighting the importance of actin dynamics regulation for the maintenance of NMJ integrity.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Ethanesulfonic acid sodium salt monohydrate, ≥98.0% (T)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)