Merck
  • Evolutionarily conserved sequence motif analysis guides development of chemically defined hydrogels for therapeutic vascularization.
Evolutionarily conserved sequence motif analysis guides development of chemically defined hydrogels for therapeutic vascularization.
Science advances (2020-09-15)
Jia Jia, Eun Je Jeon, Mei Li, Dylan J Richards, Soojin Lee, Youngmee Jung, Ryan W Barrs, Robert Coyle, Xiaoyang Li, James C Chou, Michael J Yost, Sharon Gerecht, Seung-Woo Cho, Ying Mei
ABSTRACT
Biologically active ligands (e.g., RGDS from fibronectin) play critical roles in the development of chemically defined biomaterials. However, recent decades have shown only limited progress in discovering novel extracellular matrix-protein-derived ligands for translational applications. Through motif analysis of evolutionarily conserved RGD-containing regions in laminin (LM) and peptide-functionalized hydrogel microarray screening, we identified a peptide (a1) that showed superior supports for endothelial cell (EC) functions. Mechanistic studies attributed the results to the capacity of a1 engaging both LM- and Fn-binding integrins. RNA sequencing of ECs in a1-functionalized hydrogels showed ~60% similarities with Matrigel in "vasculature development" gene ontology terms. Vasculogenesis assays revealed the capacity of a1-formulated hydrogels to improve EC network formation. Injectable alginates functionalized with a1 and MMPQK (a vascular endothelial growth factor-mimetic peptide with a matrix metalloproteinase-degradable linker) increased blood perfusion and functional recovery over decellularized extracellular matrix and (RGDS + MMPQK)-functionalized hydrogels in an ischemic hindlimb model, illustrating the power of this approach.
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse
Sigma-Aldrich
Anti-Actin Antibody, smooth muscle γ & α actin, clone CGA7, ascites fluid, clone CGA7, Chemicon®
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone 25E11, clone 25E11, Chemicon®, from mouse
Sigma-Aldrich
Anti-von Willebrand Factor Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Integrin αV Antibody, clone P3G8, clone P3G8, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, clone 6S6, Chemicon®, from mouse