- The Potential Diagnostic Utility of TROP-2 in Thyroid Neoplasms.
The Potential Diagnostic Utility of TROP-2 in Thyroid Neoplasms.
Human trophoblast cell-surface marker (TROP-2) has been reported to be overexpressed in various human carcinomas (CAs) and suggested to be a prognostic marker for some CAs. The diagnostic utility of TROP-2 in CAs has not been explored. Immunohistochemical evaluation of TROP-2 expression on tissue microarray sections of 136 thyroid neoplasms, surgical specimens of 61 atypical thyroid follicular-patterned lesions [including 33 papillary thyroid carcinomas (PTCs), 17 atypical follicular neoplasms (AFNs), and 11 adenomatoid nodules with focal nuclear atypia (ANFNA)], and 20 benign thyroid lesions, as well as 10 cytology specimens of PTCs was performed. For comparison, immunoassay for Hector Battifora mesothelial-1 (HBME-1), galectin-3, and cytokeratin 19 was performed on the 61 atypical thyroid follicular-patterned lesions. Strong membranous staining with TROP-2 was seen in 94% (33/35) of classic PTCs and 81% (30/37) of confirmed follicular variant PTCs on tissue microarray and routine surgical sections, as well as 100% (10/10) of PTCs on cytology specimens; it was not observed in follicular adenomas (n=51) or CAs (n=37), AFNs or ANFNA (n=28), benign (n=20) or normal (n=15) thyroid tissue. In contrast, the expression of HBME-1 and galectin-3 was identified in 100% (33/33) of surgical cases of PTCs and in 57% (16/28) and 50% (14/28) of AFNs and ANFNA, respectively. Our findings demonstrate that a membranous TROP-2 staining pattern is highly specific for PTC, which may serve as a potential diagnostic marker aiding in the accurate classification of morphologically equivocal thyroid follicular-patterned lesions.