- Glycosphingolipid synthesis employing a combination of recombinant glycosyltransferases and an endoglycoceramidase glycosynthase.
Glycosphingolipid synthesis employing a combination of recombinant glycosyltransferases and an endoglycoceramidase glycosynthase.
Chemical communications (Cambridge, England) (2011-09-01)
Jamie R Rich, Anna-Maria Cunningham, Michel Gilbert, Stephen G Withers
PMID21879043
ABSTRACT
Glycosynthase mutants of Rhodococcus sp. endo-glycoceramidase II efficiently synthesize complex glycosphingolipids. Glycosyl fluoride donors may be assembled via sequential glycosyltransferase-catalysed glycosylation of lactosyl fluoride. Alternatively, lactosyl fluoride may be coupled to sphingosine prior to subsequent glycosylation steps.