Merck
  • Preferential Equilibrium Partitioning of Positively Charged Tryptophan into Phosphatidylcholine Bilayer Membranes.
Preferential Equilibrium Partitioning of Positively Charged Tryptophan into Phosphatidylcholine Bilayer Membranes.
The journal of physical chemistry. B (2018-11-28)
Cari M Anderson, Alfredo Cardenas, Ron Elber, Lauren J Webb
ABSTRACT
The interactions between small molecules and lipid bilayers play a critical role in the function of cellular membranes. Understanding how a small molecule interacts with the lipid bilayer differently based on its charge reveals primordial mechanisms of transport across membranes and assists in the design of drug molecules that can penetrate cells. We have previously reported that tryptophan permeated through a phosphatidylcholine lipid bilayer membrane at a faster rate when it was positively charged (Trp+) than when negatively charged (Trp-), which corresponded to a lower potential of mean force (PMF) barrier determined through simulations. In this report, we demonstrate that Trp+ partitions into the lipid bilayer membrane to a greater degree than Trp- by interacting with the ester linkage of a phosphatidylcholine lipid, where it is stabilized by the electron withdrawing glycerol functional group. These results are in agreement with tryptophan's known role as an anchor for transmembrane proteins, though the tendency for binding of a positively charged tryptophan is surprising. We discuss the implications of our results on the mechanisms of unassisted permeation and penetration of small molecules within and across lipid bilayer membranes based on molecular charge, shape, and molecular interactions within the bilayer structure.
Product Description
Avanti
16:0-18:0(4,5-dibromo) PC, Avanti Research - A Croda Brand 850479C