Merck
  • Btk SH2-kinase interface is critical for allosteric kinase activation and its targeting inhibits B-cell neoplasms.
Btk SH2-kinase interface is critical for allosteric kinase activation and its targeting inhibits B-cell neoplasms.
Nature communications (2020-05-10)
Daniel P Duarte, Allan J Lamontanara, Giuseppina La Sala, Sukyo Jeong, Yoo-Kyoung Sohn, Alejandro Panjkovich, Sandrine Georgeon, Tim Kükenshöner, Maria J Marcaida, Florence Pojer, Marco De Vivo, Dmitri Svergun, Hak-Sung Kim, Matteo Dal Peraro, Oliver Hantschel
ABSTRACT
Bruton's tyrosine kinase (Btk) is critical for B-cell maturation and activation. Btk loss-of-function mutations cause human X-linked agammaglobulinemia (XLA). In contrast, Btk signaling sustains growth of several B-cell neoplasms which may be treated with tyrosine kinase inhibitors (TKIs). Here, we uncovered the structural mechanism by which certain XLA mutations in the SH2 domain strongly perturb Btk activation. Using a combination of molecular dynamics (MD) simulations and small-angle X-ray scattering (SAXS), we discovered an allosteric interface between the SH2 and kinase domain required for Btk activation and to which multiple XLA mutations map. As allosteric interactions provide unique targeting opportunities, we developed an engineered repebody protein binding to the SH2 domain and able to disrupt the SH2-kinase interaction. The repebody prevents activation of wild-type and TKI-resistant Btk, inhibiting Btk-dependent signaling and proliferation of malignant B-cells. Therefore, the SH2-kinase interface is critical for Btk activation and a targetable site for allosteric inhibition.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Naphthol AS-BI Phosphoric Acid Solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid