Merck
  • Imaging of the ex vivo transglutaminase activity in liver macrophages of sepsis mice.
Imaging of the ex vivo transglutaminase activity in liver macrophages of sepsis mice.
Analytical biochemistry (2020-03-07)
Ting Su, Xian-Yang Qin, Yutaka Furutani, Wenkui Yu, Soichi Kojima
ABSTRACT
Sepsis is the leading cause of death in hospitalized patients and is characterized by a dysregulated inflammatory response to infection and multiple organ failure, including the liver. Transglutaminase 2 (TG2) is a multifunctional enzyme that exhibits transamidase, GTPase, and integrin-binding activities and has opposing roles in the regulation of cell growth, differentiation, and apoptosis. TG2 plays both pathogenic and protective roles in liver diseases, revealing the need to examine the activities of TG2. Here, we introduced an ex vivo imaging approach to examine the in vivo transamidase activity of TG2 based on the combination of intraperitoneal injection of 5-biotinamidopentylamine (5BAPA), a biotinylated substrate for TG2, and fluorescent streptavidin staining in frozen liver sections. Increased 5BAPA signals was observed in the livers of lipopolysaccharide (LPS) and cecal ligation and puncture (CLP)-induced sepsis mice. Pharmacological inhibition of TG2 activity ameliorated LPS-induced liver injury. 5BAPA signals were observed in TG2-expressing and F4/80-positive midzonal macrophages, providing direct evidence that activated macrophages are the major cellular source of active TG2 in the livers of sepsis mice. Further studies focusing on the activation of 5BAPA-stained midzonal macrophages may improve understanding of the molecular pathophysiology and the development of therapeutic strategies for sepsis.
Sigma-Aldrich
Octoclothepin maleate salt, solid
Sigma-Aldrich
tert-Amylamine, 98%