Merck
  • Cyclic-AMP induces Nogo-A receptor NgR1 internalization and inhibits Nogo-A-mediated collapse of growth cone.
Cyclic-AMP induces Nogo-A receptor NgR1 internalization and inhibits Nogo-A-mediated collapse of growth cone.
Biochemical and biophysical research communications (2020-01-18)
Rayudu Gopalakrishna, Aubree Mades, Andrew Oh, Angela Zhu, Julie Nguyen, Charlotte Lin, Mark S Kindy, William J Mack
ABSTRACT
The promotion of axonal regeneration is required for functional recovery from stroke and various neuronal injuries. However, axonal regeneration is inhibited by diverse axonal growth inhibitors, such as Nogo-A. Nogo-66, a C-terminal domain of Nogo-A, binds to the Nogo-A receptor 1 (NgR1) and induces the collapse of growth cones and inhibits neurite outgrowth. NgR1 is also a receptor for additional axonal growth inhibitors, suggesting it is an important target for the prevention of axonal growth inhibition. By using the indirect immunofluorescence method, we show for the first time that a cell-permeable cAMP analog (dibutyryl-cAMP) induced a rapid decrease in the cell surface expression of NgR1 in Neuroscreen-1 (NS-1) cells. The biotinylation method revealed that cAMP indeed induced internalization of NgR1 within minutes. Other intracellular cAMP-elevating agents, such as forskolin, which directly activates adenylyl cyclase, and rolipram, which inhibits cyclic nucleotide phosphodiesterase, also induced this process. This internalization was found to be reversible and influenced by intracellular levels of cAMP. Using selective activators and inhibitors of protein kinase A (PKA) and the exchange protein directly activated by cAMP (Epac), we found that NgR1 internalization is independent of PKA, but dependent on Epac. The decrease in cell surface expression of NgR1 desensitized NS-1 cells to Nogo-66-induced growth cone collapse. Therefore, it is likely that besides axonal growth inhibitors affecting neurons, neurons themselves also self-regulate their sensitivity to axonal growth inhibitors, as influenced by intracellular cAMP/Epac. This normal cellular regulatory mechanism may be pharmacologically exploited to overcome axonal growth inhibitors, and enhance functional recovery after stroke and neuronal injuries.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human RTN4R