Merck
  • Tylophorine-based compounds are therapeutic in rheumatoid arthritis by targeting the caprin-1 ribonucleoprotein complex and inhibiting expression of associated c-Myc and HIF-1α.
Tylophorine-based compounds are therapeutic in rheumatoid arthritis by targeting the caprin-1 ribonucleoprotein complex and inhibiting expression of associated c-Myc and HIF-1α.
Pharmacological research (2019-12-04)
Yue-Zhi Lee, Huan-Chen Guo, Guan-Hao Zhao, Cheng-Wei Yang, Hsin-Yu Chang, Ruey-Bing Yang, Linyi Chen, Shiow-Ju Lee
ABSTRACT
Interruption of the Warburg effect - the observation that un-stimulated macrophages reprogram their core metabolism from oxidative phosphorylation toward aerobic glycolysis to become pro-inflammatory M1 macrophages upon stimulation - is an emerging strategy for the treatment of cancer and anti-inflammatory diseases such as rheumatoid arthritis. We studied this process with view to the discovery of novel therapeutics, and found that tylophorine-based compounds targeted a ribonucleoprotein complex containing caprin-1 and mRNAs of c-Myc and HIF-1α in LPS/IFN-γ stimulated Raw264.7 cells, diminished the protein levels of c-Myc and HIF-1α, and consequently downregulated their targeted genes that are associated with the Warburg effect, as well as the pro-inflammatory iNOS and COX2. The tylophorine-based compound DBQ 33b significantly meliorated the severity and incidence of type II collagen-monoclonal antibody-induced rheumatoid arthritis and diminished gene expressions of c-Myc, HIF-1α, iNOS, COX2, TNFα, and IL-17A in vivo. Moreover, pharmacological inhibition of either c-Myc or HIF-1α exhibited similar effects as the tylophorine-based compound DBQ 33b, even though inhibition of c-Myc reversed the induction of iNOS and COX2 in LPS/IFN-γ stimulated Raw264.7 cells to a lesser degree. Therefore, simultaneous inhibition of both c-Myc and HIF-1α is efficacious for anti-inflammation in vitro and in vivo and merits further study.
Sigma-Aldrich
KC7F2, ≥98% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
LPS, E. Coli O111:B4