Merck
  • FKBP8 LIRL-dependent mitochondrial fragmentation facilitates mitophagy under stress conditions.
FKBP8 LIRL-dependent mitochondrial fragmentation facilitates mitophagy under stress conditions.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-08)
Seung-Min Yoo, Shun-Ichi Yamashita, Hyunjoo Kim, DoHyeong Na, Haneul Lee, Seo Jin Kim, Dong-Hyung Cho, Tomotake Kanki, Yong-Keun Jung
ABSTRACT
Mitochondrial quality control maintains mitochondrial function by regulating mitochondrial dynamics and mitophagy. Despite the identification of mitochondrial quality control factors, little is known about the crucial regulators coordinating both mitochondrial fission and mitophagy. Through a cell-based functional screening assay, FK506 binding protein 8 (FKBP8) was identified to target microtubule-associated protein 1 light chain 3 (LC3) to the mitochondria and to change mitochondrial morphology. Microscopy analysis revealed that the formation of tubular and enlarged mitochondria was observed in FKBP8 knockdown HeLa cells and the cortex of Fkbp8 heterozygote-knockout mouse embryos. Under iron depletion-induced stress, FKBP8 was recruited to the site of mitochondrial division through budding and colocalized with LC3. FKBP8 was also found to be required for mitochondrial fragmentation and mitophagy under hypoxic stress. Conversely, FKBP8 overexpression induced mitochondrial fragmentation in HeLa cells, human fibroblasts and mouse embryo fibroblasts (MEFs), and this fragmentation occurred in Drp1 knockout MEF cells, FIP200 knockout HeLa cells and BNIP3/NIX double knockout HeLa cells, but not in Opa1 knockout MEFs. Interestingly, we found an LIR motif-like sequence (LIRL), as well as an LIR motif, at the N-terminus of FKBP8 and LIRL was essential for both inducing mitochondrial fragmentation and binding of FKBP8 to OPA1. Together, we suggest that FKBP8 plays an essential role in mitochondrial fragmentation through LIRL during mitophagy and this activity of FKBP8 together with LIR is required for mitophagy under stress conditions.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Polyethylenimine hydrochloride, linear, average Mn 10,000, PDI ≤1.5