Merck
  • Genome-wide CRISPR screen identifies host dependency factors for influenza A virus infection.
Genome-wide CRISPR screen identifies host dependency factors for influenza A virus infection.
Nature communications (2020-01-11)
Bo Li, Sara M Clohisey, Bing Shao Chia, Bo Wang, Ang Cui, Thomas Eisenhaure, Lawrence D Schweitzer, Paul Hoover, Nicholas J Parkinson, Aharon Nachshon, Nikki Smith, Tim Regan, David Farr, Michael U Gutmann, Syed Irfan Bukhari, Andrew Law, Maya Sangesland, Irit Gat-Viks, Paul Digard, Shobha Vasudevan, Daniel Lingwood, David H Dockrell, John G Doench, J Kenneth Baillie, Nir Hacohen
ABSTRACT
Host dependency factors that are required for influenza A virus infection may serve as therapeutic targets as the virus is less likely to bypass them under drug-mediated selection pressure. Previous attempts to identify host factors have produced largely divergent results, with few overlapping hits across different studies. Here, we perform a genome-wide CRISPR/Cas9 screen and devise a new approach, meta-analysis by information content (MAIC) to systematically combine our results with prior evidence for influenza host factors. MAIC out-performs other meta-analysis methods when using our CRISPR screen as validation data. We validate the host factors, WDR7, CCDC115 and TMEM199, demonstrating that these genes are essential for viral entry and regulation of V-type ATPase assembly. We also find that CMTR1, a human mRNA cap methyltransferase, is required for efficient viral cap snatching and regulation of a cell autonomous immune response, and provides synergistic protection with the influenza endonuclease inhibitor Xofluza.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Influenza A Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Influenza A Antibody, nucleoprotein, clone A1, FITC-conjugated, clone A1, Chemicon®, from mouse
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)