Merck
  • Total, Bioavailable, and Free Vitamin D Levels and Their Prognostic Value in Pulmonary Arterial Hypertension.
Total, Bioavailable, and Free Vitamin D Levels and Their Prognostic Value in Pulmonary Arterial Hypertension.
Journal of clinical medicine (2020-02-12)
Maria Callejo, Gema Mondejar-Parreño, Sergio Esquivel-Ruiz, Miguel A Olivencia, Laura Moreno, Isabel Blanco, Pilar Escribano-Subias, Angel Cogolludo, Joan Albert Barbera, Francisco Perez-Vizcaino
ABSTRACT
Introduction: Epidemiological studies suggest a relationship between vitamin D deficiency and cardiovascular and respiratory diseases. However, whether total, bioavailable, and/or free vitamin D levels have a prognostic role in pulmonary arterial hypertension (PAH) is unknown. We aimed to determine total, bioavailable, and free 25-hydroxy-vitamin D (25(OH)vitD) plasma levels and their prognostic value in PAH patients. Methods: In total, 67 samples of plasma from Spanish patients with idiopathic, heritable, or drug-induced PAH were obtained from the Spanish PH Biobank and compared to a cohort of 100 healthy subjects. Clinical parameters were obtained from the Spanish Registry of PAH (REHAP). Results: Seventy percent of PAH patients had severe vitamin D deficiency (total 25(OH)vitD < 10 ng/mL) and secondary hyperparathyroidism. PAH patients with total 25(OH)vitD plasma above the median of this cohort (7.17 ng/mL) had better functional class and higher 6-min walking distance and TAPSE (tricuspid annular plane systolic excursion). The main outcome measure of survival was significantly increased in these patients (age-adjusted hazard ratio: 5.40 (95% confidence interval: 2.88 to 10.12)). Vitamin D-binding protein (DBP) and albumin plasma levels were downregulated in PAH. Bioavailable 25(OH)vitD was decreased in PAH patients compared to the control cohort. Lower levels of bioavailable 25(OH)vitD (<0.91 ng/mL) were associated with more advanced functional class, lower exercise capacity, and higher risk of mortality. Free 25(OH)vitD did not change in PAH; however, lower free 25(OH)vitD (<1.53 pg/mL) values were also associated with high risk of mortality. Conclusions: Vitamin D deficiency is highly prevalent in PAH, and low levels of total 25(OH)vitD were associated with poor prognosis.
Sigma-Aldrich
BCG Albumin Assay Kit, sufficient for 250 colorimetric tests